Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166850.1 | NP_001160322.1 | 1 | 422 |
Gene Symbol: Vmn1r104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166738.1 | NP_001160210.1 | 1 | 422 |
Gene Symbol: Vmn1r107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166759.1 | NP_001160231.1 | 1 | 422 |
Gene Symbol: Vmn1r116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166744.1 | NP_001160216.1 | 1 | 422 |
Gene Symbol: Vmn1r128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166739.1 | NP_001160211.1 | 1 | 422 |
Gene Symbol: Vmn1r130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166848.1 | NP_001160320.1 | 1 | 422 |
Gene Symbol: Vmn1r135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166747.1 | NP_001160219.1 | 1 | 422 |
Gene Symbol: Vmn1r137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166849.1 | NP_001160321.1 | 1 | 422 |
Gene Symbol: Vmn1r143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166750.1 | NP_001160222.1 | 1 | 422 |
Gene Symbol: Vmn1r152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166752.1 | NP_001160224.1 | 1 | 422 |
Gene Symbol: Vmn1r155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166753.1 | NP_001160225.1 | 1 | 422 |
Gene Symbol: Vmn1r163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166755.1 | NP_001160227.1 | 1 | 422 |
Gene Symbol: Vmn1r95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167538.1 | NP_001161010.1 | 1 | 422 |