Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152810.2 | NP_690023.1 | 12, 13 | 1920 | ||
GenBank mRNA | AK004547.2 | - | 12, 13 | 1950 | ||
AK146892.1 | - | 12, 13 | 1740 | |||
AK150562.1 | - | 12, 13 | 1737 | |||
AK150861.1 | - | 12, 13 | 1737 | |||
AK172950.1 | - | 12, 13 | 1740 | |||
BC003893.1 | - | 13, 14 | 1855 | 3593535 | ||
BC031480.1 | - | 12, 13 | 1920 | 3498853 |