Search
Search

Cd99| Gene Symbol: Cd99 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025584.2 | NP_079860.2 | 822 | |||
| GenBank mRNA | AB122023.1 | - | 822 | |||
| AK002762.1 | - | 733 | ||||
| AK004342.1 | - | 824 | ||||
| AK010597.1 | - | 806 | ||||
| AK044051.1 | - | 812 | ||||
| AY262355.1 | - | 795 | ||||
| AY262356.1 | - | 790 | ||||
| BC019482.1 | - | 820 | 4218841 | |||