Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003007.4 | NP_002998.1 | 2 | 833 | ||
GenBank mRNA | BC005229.1 | - | 2 | 764 | ||
BC007096.1 | - | 2 | 764 | 4251523 | ||
BC055416.1 | - | 2 | 764 | 6668871 | ||
BT007177.1 | - | 2 | 743 | |||
J04440.1 | - | 2 | 749 |