Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asgr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012503.2 | NP_036635.1 | 5, 6 | 424 | ||
XM_006246579.1 | XP_006246641.1 | 5, 6 | 536 | |||
XM_008767768.1 | XP_008765990.1 | 5, 6 | 556 | |||
XM_008767769.1 | XP_008765991.1 | 5, 6 | 500 | |||
GenBank mRNA | BC088154.1 | - | 5, 6 | 424 | 7372227 | |
FQ209763.1 | - | 5, 6 | 460 | |||
FQ218553.1 | - | 5, 6 | 500 | |||
FQ218644.1 | - | 5, 6 | 460 | |||
FQ218824.1 | - | 5, 6 | 460 | |||
FQ219107.1 | - | 5, 6 | 497 | |||
FQ219710.1 | - | 5, 6 | 541 | |||
M21770.1 | - | 4, 5 | 326 |