Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EDN1 SIRNA Assays ›
Gene Symbol:
Edn1
Gene Name:
endothelin 1
Gene Aliases:
Et1
Chromosome Location:
Chr.17: 22137324 - 22143324 on Build Rnor_6.0
UniGene ID:
Rn.10918
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s127677
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Edn1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012548.2
NP_036680.1
3
471
GenBank mRNA
AF122903.1
-
1
17
M64711.1
-
3
466
Back To Top
More Information
Panther Classification:
Molecular Function -
peptide hormone
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
skeletal system development
prostaglandin biosynthetic process
patterning of blood vessels
response to hypoxia
in utero embryonic development
histamine secretion
regulation of systemic arterial blood pressure by endothelin
regulation of pH
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
protein kinase C-activating G-protein coupled receptor signaling pathway
cell-cell signaling
heart development
respiratory gaseous exchange
body fluid secretion
regulation of blood pressure
positive regulation of cell proliferation
dorsal/ventral pattern formation
response to ozone
multicellular organism aging
positive regulation of heart rate
positive regulation of cardiac muscle hypertrophy
positive regulation of receptor biosynthetic process
neural crest cell development
phosphatidylinositol 3-kinase signaling
response to activity
artery smooth muscle contraction
vein smooth muscle contraction
glucose transport
cell growth
regulation of vasoconstriction
sensory perception of pain
calcium-mediated signaling
peptide hormone secretion
nitric oxide transport
positive regulation of cell migration
neutrophil chemotaxis
negative regulation of cAMP biosynthetic process
phospholipase D-activating G-protein coupled receptor signaling pathway
negative regulation of cellular protein metabolic process
positive regulation of prostaglandin secretion
response to lipopolysaccharide
response to testosterone
negative regulation of smooth muscle cell apoptotic process
response to prostaglandin F
response to nicotine
intracellular signal transduction
cellular response to drug
positive regulation of urine volume
positive regulation of renal sodium excretion
response to muscle stretch
epithelial fluid transport
vasoconstriction
protein kinase C deactivation
middle ear morphogenesis
positive regulation of odontogenesis
response to drug
superoxide anion generation
rhythmic excitation
response to amino acid
positive regulation of MAP kinase activity
positive regulation of JUN kinase activity
response to leptin
positive regulation of cell size
positive regulation of mitotic nuclear division
positive regulation of transcription from RNA polymerase II promoter
positive regulation of smooth muscle contraction
positive regulation of hormone secretion
negative regulation of hormone secretion
inositol phosphate-mediated signaling
blood vessel morphogenesis
positive regulation of smooth muscle cell proliferation
cartilage development
positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
negative regulation of nitric-oxide synthase biosynthetic process
membrane depolarization
regulation of sensory perception of pain
maternal process involved in parturition
positive regulation of sarcomere organization
positive regulation of prostaglandin-endoperoxide synthase activity
cellular response to calcium ion
cellular response to interferon-gamma
cellular response to interleukin-1
cellular response to tumor necrosis factor
cellular response to peptide hormone stimulus
cellular response to glucocorticoid stimulus
cellular response to mineralocorticoid stimulus
cellular response to fatty acid
cellular response to hypoxia
response to dexamethasone
response to transforming growth factor beta
cellular response to transforming growth factor beta stimulus
positive regulation of neutrophil chemotaxis
response to salt
receptor binding
cytokine activity
hormone activity
endothelin A receptor binding
endothelin B receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0