Search
Search

ALOX5AP| Gene Symbol: ALOX5AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204406.1 | NP_001191335.1 | 2 | 477 | ||
| NM_001629.3 | NP_001620.2 | 1 | 156 | |||
| GenBank mRNA | AK314143.1 | - | 2 | 477 | ||
| BC018538.1 | - | 1 | 109 | 4342203 | ||
| BG569867.1 | - | 1 | 158 | |||