Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022176.2 | NP_071512.2 | 4 | 690 | ||
GenBank mRNA | BC158563.1 | - | 4 | 690 | 7443132 | |
FQ219971.1 | - | 4 | 706 | |||
FQ221549.1 | - | 4 | 707 | |||
FQ230053.1 | - | 4 | 708 | |||
M31155.1 | - | 4 | 649 |