Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp3ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017041.1 | NP_058737.1 | 2 | 397 | ||
XM_006233363.2 | XP_006233425.1 | 2 | 892 | |||
XM_006233364.2 | XP_006233426.1 | 2 | 157 | |||
GenBank mRNA | AY724517.1 | - | 2 | 526 | ||
D90035.1 | - | 2 | 397 | |||
FQ212355.1 | - | 2 | 741 | |||
M29275.1 | - | 2 | 397 | |||
X57115.1 | - | 2 | 460 |