Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNNT3 SIRNA Assays ›
Gene Symbol:
Tnnt3
Gene Name:
troponin T type 3 (skeletal, fast)
Gene Aliases:
Tnt
Chromosome Location:
Chr.1: 215666628 - 215683628 on Build Rnor_6.0
UniGene ID:
Rn.15488
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s128528
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tnnt3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270664.1
NP_001257593.1
9
328
NM_001270665.1
NP_001257594.1
11
361
NM_001270666.1
NP_001257595.1
11
361
NM_001270668.1
NP_001257597.1
9
328
NM_001270670.1
NP_001257599.1
10
343
NM_001270671.1
NP_001257600.1
10
343
NM_001270672.1
NP_001257601.1
9
334
NM_001270673.1
NP_001257602.1
10
346
NM_001270675.1
NP_001257604.1
9
334
NM_001270676.1
NP_001257605.1
10
346
NM_001270677.1
NP_001257606.1
8
316
NM_001270678.1
NP_001257607.1
10
349
NM_031532.2
NP_113720.1
10
349
XM_006230681.2
XP_006230743.1
9
291
XM_006230683.2
XP_006230745.1
8
279
XM_006230685.2
XP_006230747.1
7
261
XM_006230686.2
XP_006230748.1
7
261
XM_008760080.1
XP_008758302.1
12
395
XM_008760081.1
XP_008758303.1
12
395
XM_008760082.1
XP_008758304.1
11
345
XM_008760083.1
XP_008758305.1
11
342
XM_008760084.1
XP_008758306.1
10
368
XM_008760085.1
XP_008758307.1
10
333
XM_008760086.1
XP_008758308.1
9
315
GenBank mRNA
CB612473.1
-
11
310
DQ062204.1
-
10
245
DQ273678.1
-
10
245
FQ214650.1
-
10
309
FQ214722.1
-
9
296
FQ214724.1
-
10
310
FQ214766.1
-
9
290
FQ214833.1
-
10
311
FQ215030.1
-
9
290
FQ215059.1
-
9
321
FQ215106.1
-
9
290
FQ215114.1
-
9
296
FQ215252.1
-
10
308
FQ215316.1
-
9
310
FQ215319.1
-
9
290
FQ215327.1
-
11
323
FQ215374.1
-
10
336
FQ215598.1
-
11
320
FQ215602.1
-
9
323
FQ215694.1
-
9
328
FQ215723.1
-
11
362
FQ215881.1
-
11
323
FQ215885.1
-
11
311
FQ215897.1
-
10
336
FQ215955.1
-
9
290
FQ216024.1
-
6
290
FQ216051.1
-
9
290
FQ216059.1
-
11
318
FQ216097.1
-
10
304
FQ216131.1
-
9
321
FQ216208.1
-
10
308
FQ216324.1
-
9
290
FQ216367.1
-
9
290
FQ216417.1
-
10
309
FQ216467.1
-
9
290
FQ216535.1
-
10
305
FQ216561.1
-
6
290
FQ216694.1
-
10
305
FQ216711.1
-
9
327
FQ216750.1
-
6
181
FQ216763.1
-
7
281
FQ216784.1
-
9
296
FQ216866.1
-
11
323
FQ216925.1
-
1
52
FQ217147.1
-
9
289
FQ217846.1
-
11
355
FQ217900.1
-
10
336
FQ223840.1
-
9
290
FQ223999.1
-
9
290
FQ224006.1
-
9
292
FQ224345.1
-
1
21
FQ224794.1
-
9
290
FQ224813.1
-
11
323
FQ224836.1
-
9
290
FQ224839.1
-
10
308
FQ224892.1
-
10
311
FQ224966.1
-
10
294
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001163664, NM_001163669, NM_001163666, NM_001163670, NM_001163668, and others.
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
muscle contraction
regulation of muscle contraction
regulation of striated muscle contraction
metabolic process
regulation of ATPase activity
actin binding
tropomyosin binding
troponin C binding
calcium-dependent ATPase activity
troponin I binding
troponin T binding
calcium-dependent protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-2px8h:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0