Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNNT3 SIRNA Assays ›
Gene Symbol:
Tnnt3
Gene Name:
troponin T type 3 (skeletal, fast)
Gene Aliases:
Tnt
Chromosome Location:
Chr.1: 215666628 - 215683628 on Build Rnor_6.0
UniGene ID:
Rn.15488
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s128530
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tnnt3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270664.1
NP_001257593.1
7, 8
216
NM_001270665.1
NP_001257594.1
9, 10
249
NM_001270666.1
NP_001257595.1
9, 10
249
NM_001270668.1
NP_001257597.1
7, 8
216
NM_001270670.1
NP_001257599.1
8, 9
231
NM_001270671.1
NP_001257600.1
8, 9
231
NM_001270672.1
NP_001257601.1
7, 8
222
NM_001270673.1
NP_001257602.1
8, 9
234
NM_001270675.1
NP_001257604.1
7, 8
222
NM_001270676.1
NP_001257605.1
8, 9
234
NM_001270677.1
NP_001257606.1
6, 7
204
NM_001270678.1
NP_001257607.1
8, 9
237
NM_031532.2
NP_113720.1
8, 9
237
XM_006230681.2
XP_006230743.1
7, 8
179
XM_006230683.2
XP_006230745.1
6, 7
167
XM_006230685.2
XP_006230747.1
5, 6
149
XM_006230686.2
XP_006230748.1
5, 6
149
XM_008760080.1
XP_008758302.1
10, 11
283
XM_008760081.1
XP_008758303.1
10, 11
283
XM_008760082.1
XP_008758304.1
9, 10
233
XM_008760083.1
XP_008758305.1
9, 10
230
XM_008760084.1
XP_008758306.1
8, 9
256
XM_008760085.1
XP_008758307.1
8, 9
221
XM_008760086.1
XP_008758308.1
7, 8
203
GenBank mRNA
CB612473.1
-
9, 10
198
DQ062204.1
-
8, 9
133
DQ273678.1
-
8, 9
133
FQ214650.1
-
8, 9
197
FQ214722.1
-
7, 8
184
FQ214724.1
-
8, 9
198
FQ214766.1
-
7, 8
178
FQ214833.1
-
8, 9
199
FQ215030.1
-
7, 8
178
FQ215059.1
-
7, 8
209
FQ215114.1
-
7, 8
184
FQ215252.1
-
8, 9
196
FQ215316.1
-
7, 8
198
FQ215319.1
-
7, 8
178
FQ215327.1
-
9, 10
211
FQ215374.1
-
8, 9
224
FQ215598.1
-
9, 10
208
FQ215602.1
-
7, 8
211
FQ215694.1
-
7, 8
216
FQ215723.1
-
9, 10
250
FQ215881.1
-
9, 10
211
FQ215885.1
-
9, 10
199
FQ215897.1
-
8, 9
224
FQ215955.1
-
7, 8
178
FQ216051.1
-
7, 8
178
FQ216059.1
-
9, 10
206
FQ216097.1
-
8, 9
192
FQ216131.1
-
7, 8
209
FQ216208.1
-
8, 9
196
FQ216324.1
-
7, 8
178
FQ216367.1
-
7, 8
178
FQ216417.1
-
8, 9
197
FQ216467.1
-
7, 8
178
FQ216480.1
-
7, 8
184
FQ216535.1
-
8, 9
193
FQ216694.1
-
8, 9
193
FQ216711.1
-
7, 8
215
FQ216750.1
-
4, 5
69
FQ216763.1
-
5, 6
169
FQ216784.1
-
7, 8
184
FQ216866.1
-
9, 10
211
FQ217147.1
-
7, 8
177
FQ217846.1
-
9, 10
243
FQ217900.1
-
8, 9
224
FQ223840.1
-
7, 8
178
FQ223999.1
-
7, 8
178
FQ224006.1
-
7, 8
180
FQ224794.1
-
7, 8
178
FQ224813.1
-
9, 10
211
FQ224836.1
-
7, 8
178
FQ224839.1
-
8, 9
196
FQ224892.1
-
8, 9
199
FQ224966.1
-
8, 9
182
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001163664, NM_001163669, NM_001163666, NM_001163670, NM_001163668, and others.
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
muscle contraction
regulation of muscle contraction
regulation of striated muscle contraction
metabolic process
regulation of ATPase activity
actin binding
tropomyosin binding
troponin C binding
calcium-dependent ATPase activity
troponin I binding
troponin T binding
calcium-dependent protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0