Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other WNT3 SIRNA Assays ›
Gene Symbol:
Wnt3
Gene Name:
wingless-type MMTV integration site family, member 3
Gene Aliases:
INT4
Chromosome Location:
Chr.10: 91830599 - 91874836 on Build Rnor_6.0
UniGene ID:
Rn.148140
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s128595
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Wnt3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105715.1
NP_001099185.1
3
601
XM_008768262.1
XP_008766484.1
3
711
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009521, BC152304, AK043014, AK141154, AK161781, and others.
Panther Classification:
Molecular Function -
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis
regulation of cell growth
osteoblast differentiation
in utero embryonic development
mesoderm formation
heart looping
positive regulation of receptor internalization
negative regulation of heart induction by canonical Wnt signaling pathway
gamete generation
axon guidance
anterior/posterior axis specification
dorsal/ventral axis specification
anterior/posterior pattern specification
COP9 signalosome assembly
positive regulation of gene expression
Wnt signaling pathway
spinal cord association neuron differentiation
hippocampus development
cell proliferation in forebrain
Wnt signaling pathway involved in forebrain neuroblast division
dorsal/ventral neural tube patterning
hemopoiesis
neuron differentiation
extracellular matrix organization
mammary gland development
positive regulation of B cell proliferation
positive regulation of protein binding
positive regulation of peptidyl-serine phosphorylation
cellular protein localization
embryonic forelimb morphogenesis
embryonic hindlimb morphogenesis
positive regulation of catenin import into nucleus
skeletal muscle cell differentiation
post-anal tail morphogenesis
inner ear morphogenesis
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
canonical Wnt signaling pathway involved in mesenchymal stem cell differentiation
canonical Wnt signaling pathway involved in osteoblast differentiation
cell fate commitment
regulation of cell differentiation
negative regulation of fat cell differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of mesodermal cell fate specification
paraxial mesodermal cell fate commitment
anatomical structure formation involved in morphogenesis
positive regulation of collateral sprouting in absence of injury
negative regulation of axon extension involved in axon guidance
positive regulation of sequence-specific DNA binding transcription factor activity
palate development
Spemann organizer formation at the anterior end of the primitive streak
canonical Wnt signaling pathway
limb development
limb bud formation
head morphogenesis
positive regulation of protein tyrosine kinase activity
mammary gland epithelium development
positive regulation of dermatome development
canonical Wnt signaling pathway involved in cardiac muscle cell fate commitment
regulation of microtubule cytoskeleton organization
platelet aggregation
axis elongation involved in somitogenesis
positive regulation of cell-cell adhesion mediated by cadherin
positive regulation of canonical Wnt signaling pathway involved in controlling type B pancreatic cell proliferation
positive regulation of neural precursor cell proliferation
positive regulation of cardiac muscle cell differentiation
molecular_function
transcription coactivator activity
frizzled binding
frizzled-2 binding
protein domain specific binding
receptor agonist activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0