Search
Search

Cth
LOC103691744| Gene Symbol: Cth | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017074.1 | NP_058770.1 | 9 | 950 | ||
| XM_006233536.2 | XP_006233598.1 | 8 | 889 | |||
| GenBank mRNA | AB052882.2 | - | 9 | 949 | ||
| AY032875.1 | - | 9 | 949 | |||
| AY641456.1 | - | 8 | 818 | |||
| BC078869.1 | - | 9 | 963 | 7109746 | ||
| D17370.1 | - | 9 | 950 | |||
| X53460.1 | - | 838 | ||||
| Gene Symbol: LOC103691744 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761574.1 | XP_008759796.1 | 997 | |||