Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lipa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012732.3 | NP_036864.3 | 5, 6 | 821 | ||
XM_006231275.2 | XP_006231337.1 | 5, 6 | 788 | |||
XM_008760321.1 | XP_008758543.1 | 4, 5 | 684 | |||
GenBank mRNA | BC072532.1 | - | 5, 6 | 821 | 7099460 | |
S81497.1 | - | 5, 6 | 767 |