Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012881.2 | NP_037013.2 | 7 | 974 | ||
XM_008769996.1 | XP_008768218.1 | 7 | 1047 | |||
GenBank mRNA | AB001382.1 | - | 6 | 868 | ||
BC078874.1 | - | 7 | 954 | 7110677 | ||
FQ213473.1 | - | 7 | 963 | |||
FQ222192.1 | - | 7 | 990 | |||
M14656.1 | - | 7 | 947 | |||
M99252.1 | - | 7 | 983 |