Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC34A1 SIRNA Assays ›
          Gene Symbol:
          SLC34A1
          Gene Name:
          solute carrier family 34 member 1
          Gene Aliases:
          FRTS2, HCINF2, NAPI-3, NPHLOP1, NPT2, NPTIIa, SLC11, SLC17A2
          Chromosome Location:
          Chr.5: 177384431 - 177412021 on Build GRCh38
          UniGene ID:
          Hs.936
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s13072

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SLC34A1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003052.4 NP_003043.3 12 1437
          XM_005265975.1 XP_005266032.1 10 1212
          XM_017009773.1 XP_016865262.1 11 1380
          XM_017009774.1 XP_016865263.1 5 535
          GenBank mRNA AK096088.1 - 12 1416
          BC035294.1 - 10 1624
          BC050385.1 - 12 1414 5760855
          BC053349.1 - 12 1424 5186103
          L13258.1 - 12 1414

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          kidney development
          phosphate ion transport
          glycoprotein metabolic process
          response to lead ion
          cellular phosphate ion homeostasis
          response to magnesium ion
          response to estradiol
          response to vitamin A
          phosphate ion transmembrane transport
          sodium ion transmembrane transport
          response to potassium ion
          indole metabolic process
          response to drug
          cellular protein metabolic process
          sodium-dependent phosphate transport
          positive regulation of membrane potential
          response to cadmium ion
          response to mercury ion
          protein homooligomerization
          phosphate ion homeostasis
          response to growth hormone
          cellular response to metal ion
          cellular response to parathyroid hormone stimulus
          tricarboxylic acid metabolic process
          cellular response to staurosporine
          response to thyroid hormone
          dentinogenesis
          gentamycin metabolic process
          arsenate ion transmembrane transport
          positive regulation of sodium-dependent phosphate transport
          positive regulation of phosphate transmembrane transport
          inorganic phosphate transmembrane transporter activity
          sodium:phosphate symporter activity
          protein binding
          sodium-dependent phosphate transmembrane transporter activity
          PDZ domain binding
          protein complex binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-5x6th:80/100.66.78.110:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0