Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC19A1 SIRNA Assays ›
          Gene Symbol:
          SLC19A1
          Gene Name:
          solute carrier family 19 member 1
          Gene Aliases:
          CHMD, FOLT, IFC1, REFC, RFC1
          Chromosome Location:
          Chr.21: 45502517 - 45563143 on Build GRCh38
          UniGene ID:
          Hs.736903
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s13084

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SLC19A1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001205206.1 NP_001192135.1 3 1000
          NM_001205207.1 NP_001192136.1 2 749
          NM_194255.2 NP_919231.1 3 1000
          XM_005261164.3 XP_005261221.1 3 973
          XM_011529696.2 XP_011527998.1 4 1432
          XM_011529698.2 XP_011528000.1 3 1036
          XM_011529700.2 XP_011528002.1 3 1295
          XM_011529701.2 XP_011528003.1 4 1245
          XM_011529702.2 XP_011528004.1 3 959
          XM_011529703.2 XP_011528005.1 3 1027
          XM_011529704.2 XP_011528006.1 3 943
          XM_011529706.2 XP_011528008.1 2 1388
          XM_011529709.2 XP_011528011.1 3 1214
          XM_017028443.1 XP_016883932.1 3 1124
          XM_017028444.1 XP_016883933.1 4 1438
          XM_017028445.1 XP_016883934.1 4 2008
          XM_017028446.1 XP_016883935.1 3 1298
          GenBank mRNA AB209069.1 - 1 51
          AF004354.1 - 2 918
          AK303168.1 - 2 736
          AK307623.1 - 3 1004
          AK310797.1 - 3 1168
          AK313125.1 - 3 964
          BC003068.2 - 3 945 3509422
          S78996.1 - 2 847
          U15939.1 - 3 941
          U17566.1 - 3 941
          U19720.1 - 3 942

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          female pregnancy
          folic acid transport
          folic acid metabolic process
          methotrexate transport
          reduced folate transmembrane transport
          folic acid binding
          folic acid transporter activity
          reduced folate carrier activity
          methotrexate transporter activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0