siRNA Details
Gene Symbol: Rnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019182.2 | NP_062055.1 | 8 | 812 | ||
XM_008770271.1 | XP_008768493.1 | 8 | 844 | |||
XM_008770272.1 | XP_008768494.1 | 8 | 799 | |||
XM_008770273.1 | XP_008768495.1 | 8 | 932 | |||
GenBank mRNA | AF022081.1 | - | 8 | 730 | ||
AY050655.1 | - | 8 | 824 | |||
BC062024.1 | - | 8 | 812 | 5623495 | ||
FQ211855.1 | - | 7 | 694 | |||
FQ223606.1 | - | 8 | 828 |

More Information
Other Species Targeted

Panther Classification:
