siRNA Details
Gene Symbol: Rnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019182.2 | NP_062055.1 | 8 | 789 | ||
XM_008770271.1 | XP_008768493.1 | 8 | 821 | |||
XM_008770272.1 | XP_008768494.1 | 8 | 776 | |||
XM_008770273.1 | XP_008768495.1 | 8 | 909 | |||
GenBank mRNA | AF022081.1 | - | 8 | 707 | ||
AY050655.1 | - | 8 | 801 | |||
BC062024.1 | - | 8 | 789 | 5623495 | ||
FQ211855.1 | - | 7 | 671 | |||
FQ223606.1 | - | 8 | 805 |

More Information
Other Species Targeted

Panther Classification:
