Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ftl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022500.4 | NP_071945.3 | 4 | 824 | ||
GenBank mRNA | BC061525.1 | - | 4 | 828 | 6921603 | |
BC086583.1 | - | 4 | 805 | 7307718 | ||
BC088756.1 | - | 4 | 805 | 7307747 | ||
FQ209407.1 | - | 4 | 824 | |||
FQ209694.1 | - | 4 | 824 | |||
FQ209816.1 | - | 4 | 824 | |||
FQ218577.1 | - | 4 | 824 | |||
FQ219483.1 | - | 4 | 824 | |||
FQ219685.1 | - | 4 | 824 | |||
FQ219809.1 | - | 4 | 824 | |||
FQ220095.1 | - | 4 | 824 | |||
FQ220123.1 | - | 4 | 824 | |||
FQ220243.1 | - | 4 | 821 | |||
FQ220651.1 | - | 4 | 824 | |||
FQ220814.1 | - | 4 | 824 | |||
FQ220884.1 | - | 4 | 824 | |||
FQ221898.1 | - | 4 | 825 | |||
FQ222411.1 | - | 3 | 728 | |||
FQ226019.1 | - | 4 | 824 | |||
FQ226254.1 | - | 4 | 780 | |||
FQ226458.1 | - | 4 | 823 | |||
FQ226986.1 | - | 4 | 821 | |||
FQ227401.1 | - | 4 | 824 | |||
FQ227800.1 | - | 4 | 824 | |||
FQ228585.1 | - | 4 | 824 | |||
FQ229176.1 | - | 4 | 824 | |||
FQ229284.1 | - | 4 | 824 | |||
FQ229370.1 | - | 4 | 824 | |||
FQ229673.1 | - | 4 | 822 | |||
FQ229757.1 | - | 4 | 824 | |||
FQ230098.1 | - | 4 | 822 | |||
FQ230445.1 | - | 4 | 824 | |||
FQ230659.1 | - | 4 | 820 | |||
FQ231322.1 | - | 4 | 824 | |||
FQ231373.1 | - | 4 | 824 | |||
FQ231415.1 | - | 3 | 680 | |||
FQ231464.1 | - | 4 | 824 | |||
FQ231570.1 | - | 4 | 824 | |||
FQ231615.1 | - | 4 | 824 | |||
FQ231781.1 | - | 4 | 820 | |||
FQ231957.1 | - | 4 | 824 | |||
FQ232031.1 | - | 4 | 824 | |||
FQ232167.1 | - | 4 | 824 | |||
FQ232317.1 | - | 4 | 780 | |||
FQ232517.1 | - | 4 | 824 | |||
FQ233772.1 | - | 4 | 825 | |||
FQ233996.1 | - | 4 | 824 | |||
FQ234074.1 | - | 4 | 824 | |||
FQ234603.1 | - | 4 | 824 | |||
FQ234647.1 | - | 4 | 824 | |||
FQ235044.1 | - | 4 | 824 | |||
FQ235250.1 | - | 4 | 824 | |||
L01122.1 | - | 4 | 777 |
Gene Symbol: LOC100360087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729553.4 | XP_002729599.1 | 1 | 839 |