Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133286.1 | NP_579820.1 | 4, 5 | 331 | ||
XM_006231436.2 | XP_006231498.1 | 4, 5 | 523 | |||
XM_006231437.2 | XP_006231499.1 | 5, 6 | 451 | |||
XM_008760398.1 | XP_008758620.1 | 5, 6 | 418 | |||
GenBank mRNA | AB079113.1 | - | 4, 5 | 331 |