Search
Search

SMARCD2| Gene Symbol: SMARCD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098426.1 | NP_001091896.1 | 7 | 1227 | ||
| XM_005257604.2 | XP_005257661.2 | 7 | 774 | |||
| XM_017024967.1 | XP_016880456.1 | 7 | 1966 | |||
| GenBank mRNA | AK300939.1 | - | 7 | 1011 | ||
| AK303951.1 | - | 7 | 813 | |||
| AK307944.1 | - | 7 | 780 | |||
| BC136322.1 | - | 8 | 1096 | 9020309 | ||
| BC142963.1 | - | 7 | 1176 | 8860380 | ||
| BM463728.1 | - | 7 | 724 | |||
| U66618.1 | - | 7 | 1170 | |||