Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SMO SIRNA Assays ›
          Gene Symbol:
          SMO
          Gene Name:
          smoothened, frizzled class receptor
          Gene Aliases:
          FZD11, Gx, SMOH
          Chromosome Location:
          Chr.7: 129188872 - 129213548 on Build GRCh38
          UniGene ID:
          Hs.437846
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s13165

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SMO
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005631.4 NP_005622.1 8 1670
          XM_011516522.2 XP_011514824.1 9 1236
          GenBank mRNA AF071494.1 - 8 1390
          AF120103.1 - 8 1448
          AU132302.1 - 4 383
          BC009989.2 - 8 1670 4127774
          U84401.1 - 8 1390

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          vasculogenesis
          osteoblast differentiation
          in utero embryonic development
          cell fate specification
          neural crest cell migration
          heart looping
          positive regulation of neuroblast proliferation
          positive regulation of mesenchymal cell proliferation
          determination of left/right asymmetry in lateral mesoderm
          type B pancreatic cell development
          G-protein coupled receptor signaling pathway
          smoothened signaling pathway
          positive regulation of hh target transcription factor activity
          ventral midline determination
          midgut development
          anterior/posterior pattern specification
          positive regulation of gene expression
          negative regulation of gene expression
          dentate gyrus development
          cerebellar cortex morphogenesis
          thalamus development
          dorsal/ventral neural tube patterning
          smoothened signaling pathway involved in ventral spinal cord patterning
          smoothened signaling pathway involved in regulation of cerebellar granule cell precursor cell proliferation
          central nervous system neuron differentiation
          cerebral cortex development
          negative regulation of epithelial cell differentiation
          hair follicle morphogenesis
          protein localization to nucleus
          multicellular organism growth
          non-canonical Wnt signaling pathway
          positive regulation of multicellular organism growth
          positive regulation of protein import into nucleus
          odontogenesis of dentin-containing tooth
          negative regulation of apoptotic process
          negative regulation of DNA binding
          positive regulation of smoothened signaling pathway
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of organ growth
          astrocyte activation
          skeletal muscle fiber development
          forebrain morphogenesis
          homeostasis of number of cells within a tissue
          positive regulation of epithelial cell proliferation
          protein stabilization
          myoblast migration
          negative regulation of hair follicle development
          canonical Wnt signaling pathway
          detection of cell density by contact stimulus involved in contact inhibition
          atrial septum morphogenesis
          mammary gland epithelial cell differentiation
          epithelial-mesenchymal cell signaling
          somite development
          pancreas morphogenesis
          left/right axis specification
          cellular response to cholesterol
          renal system development
          mesenchymal to epithelial transition involved in metanephric renal vesicle formation
          positive regulation of branching involved in ureteric bud morphogenesis
          regulation of stem cell population maintenance
          regulation of heart morphogenesis
          G-protein coupled receptor activity
          patched binding
          protein binding
          drug binding
          Wnt-protein binding
          Wnt-activated receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0