siRNA Details
Gene Symbol: Grik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111114.1 | NP_001104584.1 | 9 | 1455 | ||
NM_001111117.1 | NP_001104587.1 | 10 | 1500 | |||
NM_017241.2 | NP_058937.1 | 10 | 1500 | |||
XM_006248053.2 | XP_006248115.1 | 9 | 1839 | |||
XM_006248054.2 | XP_006248116.1 | 10 | 1884 | |||
GenBank mRNA | M83560.1 | - | 10 | 1502 | ||
M83561.1 | - | 9 | 1457 | |||
Z11712.1 | - | 10 | 1346 | |||
Z11713.1 | - | 10 | 1346 | |||
Z11714.1 | - | 10 | 1346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -





