siRNA Details
Gene Symbol: P2ry2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017255.1 | NP_058951.1 | 3 | 1106 | ||
XM_006229757.2 | XP_006229819.1 | 3 | 889 | |||
GenBank mRNA | BC061754.1 | - | 3 | 1109 | 5599916 | |
L46865.1 | - | 3 | 1106 | |||
U09402.1 | - | 3 | 981 | |||
U56839.1 | - | 2 | 764 |
