siRNA Details
Gene Symbol: Ptgfrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019243.1 | NP_062116.1 | 3 | 556 | ||
GenBank mRNA | U26595.1 | - | 3 | 556 |

Gene Symbol: Ptgfrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019243.1 | NP_062116.1 | 3 | 556 | ||
GenBank mRNA | U26595.1 | - | 3 | 556 |