Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017269.2 | NP_058965.2 | 23 | 3865 | ||
GenBank mRNA | U40790.1 | - | 24 | 3732 |