Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017291.3 | NP_058987.2 | 7 | 821 | ||
XM_008763609.1 | XP_008761831.1 | 7 | 604 | |||
GenBank mRNA | U21070.1 | - | 7 | 778 | ||
X95579.1 | - | 7 | 821 |