siRNA Details
Gene Symbol: SNRPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003097.4 | NP_003088.1 | 6 | 740 | ||
NM_022805.3 | NP_073716.1 | 8 | 935 | |||
NM_022806.3 | NP_073717.1 | 8 | 945 | |||
NM_022807.3 | NP_073718.1 | 9 | 1080 | |||
NM_022808.3 | NP_073719.1 | 8 | 972 | |||
GenBank mRNA | AF400432.1 | - | 3 | 195 | ||
AK123119.1 | - | 4 | 1231 | |||
AK222633.1 | - | 8 | 895 | |||
BC000611.2 | - | 3 | 366 | 3347412 | ||
BC003180.1 | - | 6 | 646 | 2958068 | ||
BC018809.2 | - | 6 | 635 | |||
BC024777.1 | - | 7 | 768 | 3907606 | ||
BC025178.1 | - | 6 | 612 | 4158469 | ||
BC035280.1 | - | 10 | 1114 | |||
CR450350.1 | - | 2 | 180 | |||
J04615.1 | - | 6 | 645 | |||
KJ534946.1 | - | 2 | 180 | |||
L80004.1 | - | 8 | 924 | |||
L80005.1 | - | 9 | 1070 | |||
U41303.1 | - | 6 | 645 | |||
X15892.1 | - | 4 | 522 |
Gene Symbol: SNURF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005678.4 | NP_005669.2 | 6 | 740 | ||
GenBank mRNA | AK098563.1 | - | 6 | 644 |

More Information
Panther Classification:
Molecular Function -




