siRNA Details
Gene Symbol: Slc14a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019347.2 | NP_062220.2 | 6 | 787 | ||
NM_177962.3 | NP_808877.2 | 6 | 787 | |||
GenBank mRNA | AF031642.1 | - | 6 | 787 | ||
AF041788.1 | - | 6 | 787 | |||
AF042167.1 | - | 6 | 787 | |||
U77971.1 | - | 6 | 787 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
