Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SOX4 SIRNA Assays ›
Gene Symbol:
SOX4
Gene Name:
SRY-box 4
Gene Aliases:
EVI16
Chromosome Location:
Chr.6: 21593741 - 21598619 on Build GRCh38
UniGene ID:
Hs.643910
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s13300
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
392.00
Your Price
Online offer:
380.65
380.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SOX4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003107.2
NP_003098.1
1
1107
GenBank mRNA
BC072668.1
-
2
927
6584346
X65661.1
-
1
106
X70683.1
-
1
663
Back To Top
More Information
Panther Classification:
Molecular Function -
HMG box transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
neural tube formation
pro-B cell differentiation
mitral valve morphogenesis
cardiac ventricle formation
cardiac right ventricle morphogenesis
atrial septum primum morphogenesis
noradrenergic neuron differentiation
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
heart development
positive regulation of cell proliferation
negative regulation of cell proliferation
glial cell proliferation
spinal cord development
spinal cord motor neuron differentiation
glial cell development
positive regulation of Wnt signaling pathway
T cell differentiation
endocrine pancreas development
negative regulation of protein ubiquitination
regulation of protein stability
positive regulation of insulin secretion
somatic stem cell population maintenance
ascending aorta morphogenesis
glucose homeostasis
DNA damage response, detection of DNA damage
positive regulation of apoptotic process
positive regulation of translation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of protein export from nucleus
sympathetic nervous system development
protein stabilization
canonical Wnt signaling pathway
limb bud formation
ventricular septum morphogenesis
negative regulation of cell death
neuroepithelial cell differentiation
kidney morphogenesis
cellular response to glucose stimulus
positive regulation of canonical Wnt signaling pathway
positive regulation of N-terminal peptidyl-lysine acetylation
transcription regulatory region sequence-specific DNA binding
core promoter sequence-specific DNA binding
nucleic acid binding transcription factor activity
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription coactivator activity
transcription factor activity, sequence-specific DNA binding
protein binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0