Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021672.1 | NP_067704.1 | 2 | 475 | ||
XM_008767704.1 | XP_008765926.1 | 3 | 392 | |||
GenBank mRNA | AF099912.1 | - | 2 | 475 | ||
X81899.1 | - | 2 | 475 |