Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243998.1 | NP_001230927.1 | 3 | 123 | ||
NM_001243999.1 | NP_001230928.1 | 3 | 123 | |||
NM_001244000.1 | NP_001230929.1 | 3 | 123 | |||
NM_003121.4 | NP_003112.2 | 3 | 123 | |||
GenBank mRNA | AK225225.1 | - | 3 | 121 | ||
AK292644.1 | - | 3 | 125 | |||
AK300639.1 | - | 3 | 121 | |||
BC007921.2 | - | 3 | 102 | 4309499 | ||
DA957263.1 | - | 3 | 120 | |||
X66079.1 | - | 3 | 103 | |||
X96998.1 | - | 3 | 103 |