Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glrx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022278.1 | NP_071614.1 | 1, 2 | 246 | ||
GenBank mRNA | AF167981.3 | - | 1, 2 | 246 | ||
AF319950.1 | - | 1, 2 | 215 | |||
BC061555.1 | - | 1, 2 | 275 | 6889009 | ||
FQ224532.1 | - | 1, 2 | 282 |