Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLEC SIRNA Assays ›
Gene Symbol:
Plec
Gene Name:
plectin
Gene Aliases:
Pcn, Plec1, Pltn
Chromosome Location:
Chr.7: 117230319 - 117291473 on Build Rnor_6.0
UniGene ID:
Rn.1085
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s133908
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Plec
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164296.1
NP_001157768.1
13
1500
NM_001164297.1
NP_001157769.1
13
1435
NM_001164298.1
NP_001157770.1
13
1494
NM_001164299.1
NP_001157771.1
13
1526
NM_001164302.1
NP_001157774.1
13
1480
NM_001164303.1
NP_001157775.1
13
1433
NM_001164304.1
NP_001157776.1
13
1693
NM_001164305.1
NP_001157777.1
13
1410
NM_001164307.1
NP_001157779.1
13
1507
NM_001164308.1
NP_001157780.1
13
1567
NM_022401.2
NP_071796.2
13
1886
XM_006241870.2
XP_006241932.1
14
2097
XM_006241875.2
XP_006241937.1
14
1859
XM_006241882.2
XP_006241944.1
14
2214
XM_008765592.1
XP_008763814.1
14
2690
XM_008765593.1
XP_008763815.1
15
1874
XM_008765594.1
XP_008763816.1
14
2857
XM_008765595.1
XP_008763817.1
14
3661
XM_008765596.1
XP_008763818.1
14
1484
XM_008765597.1
XP_008763819.1
14
1607
XM_008765598.1
XP_008763820.1
14
2097
GenBank mRNA
AY480022.1
-
13
1503
AY480023.1
-
13
1438
AY480024.1
-
13
1497
AY480025.1
-
13
1529
AY480026.1
-
13
1483
AY480027.1
-
13
2030
AY480028.1
-
13
1436
AY480029.1
-
13
1581
AY480030.1
-
13
1413
AY480031.1
-
13
1510
AY480032.1
-
13
1570
X59601.1
-
13
1836
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_201388, NM_201393, NM_201389, NM_011117, NM_001163549, and others.
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
female pregnancy
response to nutrient
epithelial cell differentiation
hemidesmosome assembly
actin binding
structural constituent of cytoskeleton
cytoskeletal protein binding
structural constituent of muscle
ankyrin binding
poly(A) RNA binding
protein N-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-mjkqm:80/100.66.76.235:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0