siRNA Details
Gene Symbol: Cd151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022523.1 | NP_071968.1 | 443 | |||
XM_006230576.2 | XP_006230638.1 | 586 | ||||
GenBank mRNA | AF192547.2 | - | 443 | |||
BC072515.1 | - | 3 | 412 | 7104419 | ||
FQ222194.1 | - | 463 |
Gene Symbol: LOC100911730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749017.3 | XP_003749065.1 | 4 | 503 | ||
XM_006230598.2 | XP_006230660.1 | 4 | 586 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -




