Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BIN1 SIRNA Assays ›
          Gene Symbol:
          BIN1
          Gene Name:
          bridging integrator 1
          Gene Aliases:
          AMPH2, AMPHL, SH3P9
          Chromosome Location:
          Chr.2: 127048023 - 127107400 on Build GRCh38
          UniGene ID:
          Hs.193163
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1340

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BIN1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001320632.1 NP_001307561.1 12, 13 1442
          NM_001320633.1 NP_001307562.1 13, 14 1580
          NM_001320634.1 NP_001307563.1 11, 12 1163
          NM_001320640.1 NP_001307569.1 14, 15 1709
          NM_001320641.1 NP_001307570.1 16, 17 1856
          NM_001320642.1 NP_001307571.1 17, 18 2443
          NM_004305.3 NP_004296.1 13, 14 1532
          NM_139343.2 NP_647593.1 17, 18 1949
          NM_139344.2 NP_647594.1 16, 17 1820
          NM_139345.2 NP_647595.1 14, 15 1688
          NM_139346.2 NP_647596.1 14, 15 1661
          NM_139347.2 NP_647597.1 14, 15 1724
          NM_139348.2 NP_647598.1 13, 14 1616
          NM_139349.2 NP_647599.1 13, 14 1595
          NM_139350.2 NP_647600.1 12, 13 1487
          NM_139351.2 NP_647601.1 11, 12 1397
          XM_005263642.3 XP_005263699.1 18, 19 2067
          XM_005263643.3 XP_005263700.1 17, 18 1974
          XM_005263645.2 XP_005263702.1 17, 18 1458
          XM_006712425.3 XP_006712488.1 16, 17 1935
          XM_006712426.3 XP_006712489.1 16, 17 1935
          XM_006712427.2 XP_006712490.1 16, 17 1413
          XM_006712428.3 XP_006712491.1 15, 16 1827
          XM_006712429.3 XP_006712492.1 15, 16 1806
          XM_006712431.3 XP_006712494.1 14, 15 1737
          XM_006712432.3 XP_006712495.1 14, 15 1698
          XM_006712433.3 XP_006712496.1 13, 14 1608
          XM_006712434.3 XP_006712497.1 12, 13 1563
          XM_011510975.2 XP_011509277.1 18, 19 2700
          XM_017003819.1 XP_016859308.1 18, 19 1881
          XM_017003820.1 XP_016859309.1 14, 15 1797
          XM_017003821.1 XP_016859310.1 13, 14 1692
          XM_017003822.1 XP_016859311.1 12, 13 1599
          XM_017003823.1 XP_016859312.1 13, 14 1152
          XM_017003824.1 XP_016859313.1 12, 13 1083
          XM_017003825.1 XP_016859314.1 12, 13 1377
          XM_017003826.1 XP_016859315.1 12, 13 1044
          GenBank mRNA AF001383.1 - 13, 14 1403
          AF004015.1 - 17, 18 1565
          AF043898.1 - 14, 15 1340
          AF043899.1 - 14, 15 1304
          AF043900.1 - 13, 14 1211
          AF043901.1 - 16, 17 1436
          AF068914.1 - 12, 13 1163
          AF068915.1 - 11, 12 1073
          AF068916.1 - 12, 13 1118
          AF068917.1 - 13, 14 1292
          AF068918.1 - 14, 15 1337
          AF070576.1 - 13, 14 1171
          AK127325.1 - 3, 4 4432
          AK295300.1 - 13, 14 1298
          AK301153.1 - 11, 12 1163
          AL713697.1 - 17, 18 1910
          BC004101.2 - 12, 13 1163 3688734
          BM805510.1 - 3, 4 287
          BT006865.1 - 12, 13 1103
          DR761993.1 - 5, 6 556
          U68485.1 - 13, 14 1194
          U87558.1 - 17, 18 1586

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          endocytosis
          nucleus organization
          cell proliferation
          viral process
          regulation of endocytosis
          muscle cell differentiation
          positive regulation of apoptotic process
          regulation of neuron differentiation
          positive regulation of astrocyte differentiation
          lipid tube assembly
          regulation of cell cycle arrest
          protein binding
          identical protein binding
          tau protein binding
          actin filament binding
          RNA polymerase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-gpcss:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0