Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031112.1 | NP_112374.1 | 3 | 258 | ||
XM_006252530.2 | XP_006252592.1 | 3 | 329 | |||
XM_006252531.2 | XP_006252593.1 | 3 | 329 | |||
XM_006252532.2 | XP_006252594.1 | 3 | 329 | |||
XM_006252533.2 | XP_006252595.1 | 3 | 329 | |||
GenBank mRNA | BC058140.1 | - | 3 | 269 | 6919886 | |
FQ209499.1 | - | 1 | 32 | |||
FQ221116.1 | - | 1 | 32 | |||
FQ221331.1 | - | 3 | 263 | |||
FQ221519.1 | - | 1 | 32 | |||
FQ221551.1 | - | 1 | 32 | |||
FQ221824.1 | - | 1 | 32 | |||
FQ222238.1 | - | 3 | 263 | |||
FQ222347.1 | - | 1 | 32 | |||
FQ222799.1 | - | 1 | 32 | |||
FQ222961.1 | - | 1 | 32 | |||
FQ223039.1 | - | 3 | 263 | |||
FQ223185.1 | - | 1 | 43 | |||
FQ225772.1 | - | 3 | 263 | |||
FQ228365.1 | - | 1 | 32 | |||
FQ228596.1 | - | 1 | 32 | |||
FQ228664.1 | - | 1 | 32 | |||
FQ228683.1 | - | 1 | 32 | |||
FQ229436.1 | - | 1 | 32 | |||
X51538.1 | - | 3 | 258 | |||
X52445.1 | - | 3 | 258 |