Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031131.1 | NP_112393.1 | 5 | 1088 | ||
XM_006250448.2 | XP_006250510.1 | 4 | 1914 | |||
GenBank mRNA | AF135598.1 | - | 4 | 698 | ||
AF153012.2 | - | 5 | 1088 | |||
AF153013.1 | - | 4 | 1004 | |||
AJ132718.1 | - | 4 | 712 |