Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SULT1A1 SIRNA Assays ›
          Gene Symbol:
          SULT1A1 SULT1A2
          Gene Name:
          sulfotransferase family 1A member 1
          sulfotransferase family 1A member 2
          Gene Aliases:
          HAST1/HAST2, P-PST, PST, ST1A1, ST1A3, STP, STP1, TSPST1
          HAST4, P-PST, ST1A2, STP2, TSPST2
          Chromosome Location:
          Chr.16: 28605587 - 28623586 on Build GRCh38
          Chromosome Location:
          Chr.16: 28591943 - 28597079 on Build GRCh38
          UniGene ID:
          Hs.567342
          Hs.546304
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s13585

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SULT1A1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001055.3 NP_001046.2 3 363
          NM_177529.2 NP_803565.1 3 394
          NM_177530.2 NP_803566.1 3 519
          NM_177534.2 NP_803878.1 2 718
          XM_017023604.1 XP_016879093.1 4 448
          XM_017023605.1 XP_016879094.1 4 445
          XM_017023606.1 XP_016879095.1 4 448
          XM_017023607.1 XP_016879096.1 6 1009
          XM_017023608.1 XP_016879097.1 3 363
          XM_017023609.1 XP_016879098.1 3 413
          XM_017023610.1 XP_016879099.1 3 444
          XM_017023611.1 XP_016879100.1 5 936
          XM_017023612.1 XP_016879101.1 6 860
          XM_017023613.1 XP_016879102.1 4 675
          GenBank mRNA AB062428.1 - 2 301
          AB209149.1 - 5775
          AJ007418.1 - 2 262
          BC000923.2 - 3 313 3447900
          BC110887.1 - 6 776 5190558
          BG482944.1 - 3 364
          BM927749.1 - 3 340
          BT007324.1 - 2 245
          CV023878.1 - 2 245
          L10819.1 - 3 283
          L19955.1 - 2 286
          L19999.1 - 3 315
          U09031.1 - 2 286
          U26309.1 - 2 310
          X78283.1 - 3 404
          X84654.1 - 2 259
          Gene Symbol: SULT1A2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001054.3 NP_001045.1 3 323
          NM_177528.2 NP_803564.1 3 596
          XM_006721077.3 XP_006721140.1 2 959
          XM_017023600.1 XP_016879089.1 3 332
          XM_017023601.1 XP_016879090.1 3 760
          XM_017023602.1 XP_016879091.1 4 462
          XM_017023603.1 XP_016879092.1 3 328
          GenBank mRNA BC113727.1 - 3 318 8322779
          BC113729.1 - 3 318 8322781
          U28169.1 - 3 318
          U28170.1 - 3 306
          X78282.1 - 2 245

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          catecholamine metabolic process
          xenobiotic metabolic process
          steroid metabolic process
          estrogen metabolic process
          amine metabolic process
          flavonoid metabolic process
          3'-phosphoadenosine 5'-phosphosulfate metabolic process
          sulfation
          amine biosynthetic process
          phenol-containing compound metabolic process
          aryl sulfotransferase activity
          protein binding
          sulfotransferase activity
          flavonol 3-sulfotransferase activity
          steroid sulfotransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0