Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS2 SIRNA Assays ›
          Gene Symbol:
          Rps2 LOC100125366
          Gene Name:
          ribosomal protein S2
          40S ribosomal protein S2
          Gene Aliases:
          Rps2r1
          -
          Chromosome Location:
          Chr.10: 14088171 - 14090020 on Build Rnor_6.0
          Chromosome Location:
          Chr.5: 166864301 - 166872127 on Build Rnor_6.0
          UniGene ID:
          Rn.2115
          Rn.2115
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s136258

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rps2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031838.3 NP_114026.3 4 498
          GenBank mRNA BC098726.1 - 4 487 7378399
          CB324420.1 - 5 517
          CD372915.1 - 4 498
          FQ209478.1 - 5 509
          FQ210378.1 - 5 510
          FQ213061.1 - 5 510
          FQ213116.1 - 5 507
          FQ214025.1 - 5 510
          FQ215811.1 - 5 512
          FQ216756.1 - 5 510
          FQ218484.1 - 5 510
          FQ218550.1 - 5 511
          FQ218641.1 - 5 509
          FQ218643.1 - 5 513
          FQ219176.1 - 5 509
          FQ219789.1 - 5 510
          FQ219844.1 - 5 512
          FQ219893.1 - 5 509
          FQ220315.1 - 5 510
          FQ220515.1 - 5 510
          FQ220642.1 - 5 510
          FQ220831.1 - 5 509
          FQ220841.1 - 5 510
          FQ220885.1 - 5 510
          FQ220902.1 - 5 510
          FQ221771.1 - 5 510
          FQ221977.1 - 5 509
          FQ222558.1 - 5 510
          FQ222607.1 - 5 509
          FQ222729.1 - 5 510
          FQ222900.1 - 5 510
          FQ223242.1 - 5 510
          FQ225100.1 - 6 383
          FQ225247.1 - 5 503
          FQ225937.1 - 5 507
          FQ226404.1 - 5 510
          FQ226432.1 - 5 512
          FQ226962.1 - 5 510
          FQ228202.1 - 5 510
          FQ229347.1 - 5 510
          FQ229368.1 - 5 510
          FQ229383.1 - 5 508
          FQ229400.1 - 5 509
          FQ229467.1 - 5 512
          FQ229773.1 - 4 507
          FQ229778.1 - 5 510
          FQ230085.1 - 5 509
          FQ230109.1 - 5 512
          FQ230137.1 - 5 510
          FQ230697.1 - 5 507
          FQ231866.1 - 5 510
          FQ233653.1 - 5 510
          FQ234367.1 - 5 510
          FQ234813.1 - 5 510
          U92696.1 - 5 391
          U92698.1 - 5 416
          U92699.1 - 5 390
          U92700.1 - 5 363
          X57432.1 - 5 508
          Gene Symbol: LOC100125366
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC060584.1 - 2 701 6886957

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ribosomal small subunit assembly
          formation of translation preinitiation complex
          positive regulation of transferase activity
          cellular response to interleukin-4
          RNA binding
          mRNA binding
          structural constituent of ribosome
          fibroblast growth factor binding
          enzyme binding
          protein complex binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-72rkq:80/100.66.79.224:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline