Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUPT4H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003168.2 | NP_003159.1 | 2 | 201 | ||
NR_073470.1 | 2 | 203 | ||||
GenBank mRNA | AK129758.1 | - | 2 | 172 | ||
AK311986.1 | - | 2 | 114 | |||
BC002802.2 | - | 2 | 159 | 3636217 | ||
BP196741.1 | - | 2 | 201 | |||
BP318625.1 | - | 2 | 164 | |||
BX331655.2 | - | 2 | 165 | |||
CR407663.1 | - | 2 | 98 | |||
U38817.1 | - | 2 | 140 | |||
U38818.1 | - | 2 | 159 | |||
U43923.1 | - | 2 | 165 |