Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3kbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053360.2 | NP_445812.1 | 15 | 1859 | ||
XM_006256924.2 | XP_006256986.2 | 15 | 1801 | |||
XM_006256929.2 | XP_006256991.2 | 13 | 1627 | |||
XM_006256930.2 | XP_006256992.1 | 10 | 1220 | |||
XM_006256931.1 | XP_006256993.1 | 17 | 2152 | |||
XM_008773199.1 | XP_008771421.1 | 4 | 410 | |||
XM_008773200.1 | XP_008771422.1 | 4 | 350 | |||
XM_008773201.1 | XP_008771423.1 | 4 | 306 | |||
GenBank mRNA | AF131867.1 | - | 13 | 1415 | ||
AF230519.1 | - | 15 | 1671 | |||
AF230520.1 | - | 15 | 1525 | |||
AF255885.1 | - | 3 | 577 | |||
AF255886.1 | - | 11 | 1096 | |||
AF255887.1 | - | 10 | 990 | |||
AF255888.1 | - | 15 | 1777 | |||
BC070877.1 | - | 15 | 1859 | 7096160 |