Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TAC1 SIRNA Assays ›
          Gene Symbol:
          TAC1
          Gene Name:
          tachykinin precursor 1
          Gene Aliases:
          Hs.2563, NK2, NKNA, NPK, TAC2
          Chromosome Location:
          Chr.7: 97731959 - 97740472 on Build GRCh38
          UniGene ID:
          Hs.2563
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s13707

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TAC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003182.2 NP_003173.1 3 435
          NM_013996.2 NP_054702.1 3 435
          NM_013997.2 NP_054703.1 3 435
          NM_013998.2 NP_054704.1 3 435
          GenBank mRNA AF050655.1 - 2 84
          AF050656.1 - 2 84
          AF050657.1 - 2 84
          AF050658.1 - 2 84
          AK222957.1 - 3 310
          BC018047.1 - 3 309 4792746
          CR407602.1 - 2 189
          CR541730.1 - 2 189
          DN992454.1 - 3 443
          M68906.1 - 2 85
          M68907.1 - 2 85
          U37529.1 - 3 334
          X54469.1 - 3 270

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001124770, NM_012666, NM_001124768, NM_001124769, XM_008762693, and others.
          This siRNA also targets the following transcripts in Mouse: NM_009311, BC119122, M68908, M68909, D17584, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of acute inflammatory response
          inflammatory response
          positive regulation of cytosolic calcium ion concentration
          tachykinin receptor signaling pathway
          neuropeptide signaling pathway
          cell-cell signaling
          chemical synaptic transmission
          insemination
          long-term memory
          regulation of blood pressure
          associative learning
          detection of abiotic stimulus
          response to hormone
          negative regulation of heart rate
          positive regulation of epithelial cell migration
          sensory perception of pain
          positive regulation of synaptic transmission, cholinergic
          positive regulation of synaptic transmission, GABAergic
          response to lipopolysaccharide
          positive regulation of renal sodium excretion
          response to morphine
          positive regulation of action potential
          positive regulation of ossification
          positive regulation of saliva secretion
          response to pain
          positive regulation of lymphocyte proliferation
          positive regulation of stress fiber assembly
          cellular response to nerve growth factor stimulus
          positive regulation of corticosterone secretion
          receptor binding
          substance P receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0