Search
Search

Pafah1b3| Gene Symbol: Pafah1b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053654.2 | NP_446106.1 | 6 | 580 | ||
| XM_006228365.2 | XP_006228427.1 | 6 | 718 | |||
| GenBank mRNA | AF016047.1 | - | 6 | 580 | ||
| BC126079.1 | - | 5 | 1146 | |||