Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsto1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007602.1 | NP_001007603.1 | 5 | 596 | ||
GenBank mRNA | AB008807.1 | - | 5 | 578 | ||
BC079363.1 | - | 5 | 596 | 7109248 | ||
FJ179405.1 | - | 5 | 514 | |||
FQ218105.1 | - | 5 | 571 | |||
FQ220742.1 | - | 5 | 538 |