Search
Search

TARS| Gene Symbol: TARS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258437.1 | NP_001245366.1 | 15 | 1724 | ||
| NM_001258438.1 | NP_001245367.1 | 15 | 1997 | |||
| NM_152295.4 | NP_689508.3 | 14 | 1898 | |||
| NR_047676.1 | 15 | 2019 | ||||
| NR_047677.1 | 13 | 1737 | ||||
| NR_047678.1 | 13 | 1707 | ||||
| GenBank mRNA | AK095852.1 | - | 13 | 1519 | ||
| AK222804.1 | - | 14 | 1655 | |||
| AK292346.1 | - | 15 | 1724 | |||
| AK293620.1 | - | 15 | 1808 | |||
| AK296783.1 | - | 13 | 1536 | |||
| AK308342.1 | - | 15 | 1803 | |||
| AK315713.1 | - | 14 | 1709 | |||
| BC000517.2 | - | 14 | 1667 | 2822851 | ||
| BC010578.1 | - | 14 | 1685 | 3458845 | ||
| BC088355.1 | - | 14 | 1676 | |||
| M63180.1 | - | 14 | 1689 | |||