Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Enterprise Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TAZ SIRNA Assays ›
Gene Symbol:
TAZ
Gene Name:
tafazzin
Gene Aliases:
BTHS, CMD3A, EFE, EFE2, G4.5, LVNCX, Taz1
Chromosome Location:
Chr.X: 154411406 - 154421726 on Build GRCh38
UniGene ID:
Hs.409911
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s13807
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TAZ
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000116.4
NP_000107.1
11
1159
NM_001303465.1
NP_001290394.1
10
1171
NM_181311.3
NP_851828.1
10
1069
NM_181312.3
NP_851829.1
10
1117
NM_181313.3
NP_851830.1
9
1027
NR_024048.2
10
1175
XM_006724836.1
XP_006724899.1
11
1190
XM_006724837.1
XP_006724900.1
8
1175
XM_006724839.1
XP_006724902.1
9
1058
XM_006724841.3
XP_006724904.1
11
911
XM_006724842.3
XP_006724905.1
10
821
XM_011531189.1
XP_011529491.1
8
977
XM_011531191.2
XP_011529493.1
10
1349
XM_017029761.1
XP_016885250.1
8
1121
XM_017029762.1
XP_016885251.1
10
1100
XM_017029763.1
XP_016885252.1
8
923
XM_017029764.1
XP_016885253.1
9
688
XM_017029765.1
XP_016885254.1
9
1226
GenBank mRNA
AK130900.1
-
5
1522
AK291848.1
-
11
1027
AY231461.1
-
10
751
AY231462.1
-
10
799
AY231463.1
-
9
709
AY231464.1
-
11
841
AY258036.1
-
11
841
AY258037.1
-
9
709
AY258038.1
-
10
799
AY258039.1
-
10
751
BC005062.1
-
10
1132
BC011515.1
-
7
840
4339983
CA488175.1
-
9
838
DQ884408.1
-
10
845
KU178348.1
-
7
743
KU178349.1
-
11
833
KU178350.1
-
10
791
KU178351.1
-
9
788
KU178352.1
-
10
743
KU178353.1
-
9
701
X92762.1
-
11
1121
Back To Top
More Information
Panther Classification:
Molecular Function -
acyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
heart development
skeletal muscle tissue development
hemopoiesis
cardiolipin biosynthetic process
mitochondrial respiratory chain complex I assembly
cardiolipin acyl-chain remodeling
cristae formation
mitochondrial ATP synthesis coupled electron transport
cardiac muscle tissue development
cardiac muscle contraction
1-acylglycerol-3-phosphate O-acyltransferase activity
O-acyltransferase activity
1-acylglycerophosphocholine O-acyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0