Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RPL6 SIRNA Assays ›
Gene Symbol:
Rpl6
Gene Name:
ribosomal protein L6
Gene Aliases:
-
Chromosome Location:
Chr.12: 40877578 - 40882030 on Build Rnor_6.0
UniGene ID:
Rn.234072
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s138477
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rpl6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_053971.3
NP_446423.2
2
354
XM_006249377.1
XP_006249439.1
3
373
XM_006249378.2
XP_006249440.1
3
407
GenBank mRNA
BC061784.1
-
2
350
BC078761.1
-
2
357
7123086
BC097295.1
-
2
358
7446680
FQ209419.1
-
1
32
FQ213055.1
-
1
32
FQ213078.1
-
1
32
FQ216758.1
-
1
32
FQ217744.1
-
1
32
FQ219956.1
-
1
32
FQ219982.1
-
1
32
FQ220180.1
-
1
32
FQ220346.1
-
1
32
FQ220362.1
-
1
32
FQ220471.1
-
1
32
FQ220678.1
-
1
32
FQ220699.1
-
1
32
FQ220887.1
-
1
32
FQ220959.1
-
1
32
FQ221615.1
-
1
32
FQ222080.1
-
1
32
FQ222331.1
-
1
32
FQ222465.1
-
1
32
FQ223301.1
-
1
32
FQ226632.1
-
1
32
FQ227060.1
-
1
32
FQ227100.1
-
1
32
FQ227955.1
-
1
32
FQ227974.1
-
1
32
FQ228583.1
-
1
32
FQ228665.1
-
1
32
FQ229345.1
-
1
32
FQ229355.1
-
1
32
FQ229484.1
-
3
372
FQ229571.1
-
1
32
FQ229624.1
-
1
32
FQ229675.1
-
1
32
FQ229700.1
-
1
32
FQ229820.1
-
1
32
FQ229854.1
-
1
32
FQ229857.1
-
1
32
FQ229943.1
-
3
371
FQ230171.1
-
1
32
FQ230222.1
-
1
32
FQ232778.1
-
1
32
FQ233198.1
-
1
32
FQ233475.1
-
1
32
FQ234880.1
-
3
371
FQ235095.1
-
1
32
X87107.1
-
3
368
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_011290, AK167439, AK151022, AK002648, AK088472, and others.
Panther Classification:
Molecular Function -
ribosomal protein
Gene Ontology Categories:
Function(s)
Process(es)
translation
structural constituent of ribosome
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-zgjsk:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0