Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS15A SIRNA Assays ›
          Gene Symbol:
          Rps15a
          Gene Name:
          ribosomal protein S15a
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 187759855 - 187766734 on Build Rnor_6.0
          UniGene ID:
          Rn.2890
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s138504

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rps15a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_053982.1 NP_446434.1 1 95
          XM_006230097.1 XP_006230159.1 2 142
          XM_006230098.1 XP_006230160.1 2 185
          GenBank mRNA BC058452.1 - 2 120 6921126
          FQ210209.1 - 2 117
          FQ211215.1 - 2 120
          FQ212042.1 - 2 116
          FQ212257.1 - 2 114
          FQ214481.1 - 2 116
          FQ220927.1 - 2 112
          FQ221112.1 - 2 124
          FQ221144.1 - 2 147
          FQ221174.1 - 2 114
          FQ221230.1 - 2 189
          FQ221308.1 - 2 118
          FQ221419.1 - 2 116
          FQ221518.1 - 2 116
          FQ221677.1 - 2 114
          FQ221720.1 - 2 185
          FQ221825.1 - 2 116
          FQ221840.1 - 2 116
          FQ221877.1 - 2 116
          FQ222252.1 - 2 127
          FQ222272.1 - 2 116
          FQ222274.1 - 2 116
          FQ222304.1 - 2 116
          FQ222376.1 - 2 116
          FQ222415.1 - 2 114
          FQ222418.1 - 2 118
          FQ222718.1 - 2 116
          FQ222721.1 - 2 114
          FQ222768.1 - 2 114
          FQ222892.1 - 2 117
          FQ222921.1 - 2 119
          FQ222922.1 - 2 185
          FQ223006.1 - 2 116
          FQ223095.1 - 2 116
          FQ223230.1 - 2 115
          FQ223341.1 - 2 116
          FQ223389.1 - 2 116
          FQ223433.1 - 2 116
          FQ223787.1 - 2 116
          FQ223822.1 - 2 115
          FQ223930.1 - 238
          FQ224201.1 - 2 116
          FQ224347.1 - 2 116
          FQ224383.1 - 2 116
          FQ228419.1 - 2 115
          FQ228429.1 - 2 116
          FQ228446.1 - 2 116
          FQ228475.1 - 2 117
          FQ228508.1 - 2 116
          FQ228609.1 - 2 116
          FQ228617.1 - 2 115
          FQ229004.1 - 2 117
          FQ229042.1 - 2 114
          FQ229063.1 - 2 115
          X77953.1 - 1 95

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          positive regulation of cell proliferation
          response to virus
          positive regulation of cell cycle
          structural constituent of ribosome
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-hp62d:80/100.66.78.139:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0